siRNA Details
| Gene Symbol: 4921536K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026150.3 | NP_080426.1 | 1 | 160 | ||
| GenBank mRNA | AK006513.1 | - | 1 | 133 | ||
| AK015005.1 | - | 1 | 133 | |||
| AK049235.1 | - | 1 | 132 | |||
| AK081396.1 | - | 1 | 148 | |||
| BC048445.1 | - | 1 | 129 | 6704927 | ||

4921536K21Rik
Genomic Map