siRNA Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026162.6 | NP_080438.2 | 3 | 1213 | ||
| GenBank mRNA | AF378761.1 | - | 3 | 412 | ||
| AK004640.1 | - | 3 | 1195 | |||
| AK019482.2 | - | 3 | 1169 | |||
| AK035165.1 | - | 3 | 1214 | |||
| AK154921.1 | - | 3 | 1160 | |||
| BC057881.1 | - | 3 | 473 | 5349869 | ||

Plxdc2
Genomic Map