siRNA Details
Gene Symbol: Plxdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026162.6 | NP_080438.2 | 1 | 864 | ||
GenBank mRNA | AF378761.1 | - | 1 | 63 | ||
AK004640.1 | - | 1 | 846 | |||
AK019482.2 | - | 1 | 820 | |||
AK035165.1 | - | 1 | 865 | |||
AK154921.1 | - | 1 | 811 | |||
BC057881.1 | - | 1 | 124 | 5349869 |
