siRNA Details
Gene Symbol: Gpalpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026177.3 | NP_080453.2 | 8 | 1043 | ||
GenBank mRNA | AK004707.1 | - | 8 | 1023 | ||
AK016681.1 | - | 8 | 1050 | |||
AK082912.1 | - | 8 | 1024 | |||
AK173238.1 | - | 8 | 988 | |||
BC029731.1 | - | 8 | 1004 | 4946215 |
