siRNA Details
| Gene Symbol: Cwc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026186.4 | NP_080462.2 | 8, 9 | 1133 | ||
| GenBank mRNA | AK004983.2 | - | 8, 9 | 1077 | ||
| AK017849.1 | - | 8, 9 | 1073 | |||
| AK035158.1 | - | 8, 9 | 1061 | |||
| BC021478.1 | - | 1047 | ||||
| BC053447.1 | - | 1, 2 | 287 | |||

Cwc25
Genomic Map