siRNA Details
| Gene Symbol: Tmem167b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026198.2 | NP_080474.2 | 3 | 368 | ||
| GenBank mRNA | AK008415.1 | - | 3 | 281 | ||
| AK155399.1 | - | 3 | 264 | |||
| BC050932.1 | - | 3 | 272 | 6811263 | ||
| BC055449.1 | - | 3 | 285 | 3993993 | ||
| BQ560740.1 | - | 3 | 368 | |||

Tmem167b
Genomic Map