siRNA Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025615.3 | NP_001020786.1 | 3, 4 | 383 | ||
| NM_001289436.1 | NP_001276365.1 | 4, 5 | 429 | |||
| NM_026202.4 | NP_080478.2 | 3, 4 | 383 | |||
| GenBank mRNA | AJ534985.1 | - | 3, 4 | 250 | ||
| AK012190.1 | - | 3, 4 | 277 | |||
| AK016827.1 | - | 3, 4 | 358 | |||
| AK077557.1 | - | 3, 4 | 321 | |||
| AK147820.1 | - | 3, 4 | 334 | |||
| AK151672.1 | - | 3, 4 | 345 | |||
| AK162434.1 | - | 3, 4 | 360 | |||
| AK165214.1 | - | 3, 4 | 288 | |||
| BC144895.1 | - | 3, 4 | 362 | 9053443 | ||
| BC144898.1 | - | 4, 5 | 408 | |||
| BC145933.1 | - | 3, 4 | 362 | 40131236 | ||
| BC145935.1 | - | 3, 4 | 362 | 40131238 | ||
| BY716501.1 | - | 3, 4 | 358 | |||
| CJ045424.1 | - | 3, 4 | 387 | |||

Ccdc50
Genomic Map