siRNA Details
| Gene Symbol: Rnf151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026205.3 | NP_080481.1 | 2, 3 | 211 | ||
| GenBank mRNA | AK005850.1 | - | 2, 3 | 172 | ||
| AK005854.1 | - | 2, 3 | 167 | |||
| BC049562.1 | - | 2, 3 | 211 | 6774166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf151| Gene Symbol: Rnf151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026205.3 | NP_080481.1 | 2, 3 | 211 | ||
| GenBank mRNA | AK005850.1 | - | 2, 3 | 172 | ||
| AK005854.1 | - | 2, 3 | 167 | |||
| BC049562.1 | - | 2, 3 | 211 | 6774166 | ||