siRNA Details
| Gene Symbol: Saysd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026209.1 | NP_080485.1 | 2 | 327 | ||
| GenBank mRNA | AK007941.1 | - | 282 | |||
| AK031748.1 | - | 2 | 253 | |||
| BC027644.1 | - | 2 | 248 | 1248085 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saysd1| Gene Symbol: Saysd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026209.1 | NP_080485.1 | 2 | 327 | ||
| GenBank mRNA | AK007941.1 | - | 282 | |||
| AK031748.1 | - | 2 | 253 | |||
| BC027644.1 | - | 2 | 248 | 1248085 | ||