siRNA Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014974.1 | NP_001014974.1 | 3 | 1367 | ||
| GenBank mRNA | AK129075.1 | - | 1 | 194 | ||
| AM690747.1 | - | 1 | 970 | |||
| BC085151.1 | - | 3 | 1367 | 30534216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll4| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014974.1 | NP_001014974.1 | 3 | 1367 | ||
| GenBank mRNA | AK129075.1 | - | 1 | 194 | ||
| AM690747.1 | - | 1 | 970 | |||
| BC085151.1 | - | 3 | 1367 | 30534216 | ||