siRNA Details
| Gene Symbol: Zswim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178375.2 | NP_848462.1 | 2 | 1759 | ||
| GenBank mRNA | AK014904.1 | - | 2 | 1780 | ||
| BC023287.1 | - | 2 | 1759 | 3993352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim3| Gene Symbol: Zswim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178375.2 | NP_848462.1 | 2 | 1759 | ||
| GenBank mRNA | AK014904.1 | - | 2 | 1780 | ||
| BC023287.1 | - | 2 | 1759 | 3993352 | ||