siRNA Details
| Gene Symbol: Tmem35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026239.2 | NP_080515.1 | 1, 2 | 394 | ||
| GenBank mRNA | AK005180.1 | - | 1, 2 | 229 | ||
| AK018535.1 | - | 1, 2 | 231 | |||
| AK042538.1 | - | 1, 2 | 230 | |||
| AK043881.1 | - | 1, 2 | 229 | |||
| BC042623.1 | - | 1, 2 | 394 | 5369317 | ||
| BC050880.1 | - | 1, 2 | 296 | 5690508 | ||

Tmem35
Genomic Map