siRNA Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026251.2 | NP_080527.2 | 6 | 759 | ||
| GenBank mRNA | AK015195.1 | - | 6 | 760 | ||
| BC145646.1 | - | 4 | 537 | 40130949 | ||
| DV658440.1 | - | 4 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Patl2| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026251.2 | NP_080527.2 | 6 | 759 | ||
| GenBank mRNA | AK015195.1 | - | 6 | 760 | ||
| BC145646.1 | - | 4 | 537 | 40130949 | ||
| DV658440.1 | - | 4 | 370 | |||