siRNA Details
| Gene Symbol: Zfp605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163996.1 | NP_001157468.1 | 5 | 2079 | ||
| GenBank mRNA | AK134509.1 | - | 1 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp605| Gene Symbol: Zfp605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163996.1 | NP_001157468.1 | 5 | 2079 | ||
| GenBank mRNA | AK134509.1 | - | 1 | 135 | ||