siRNA Details
| Gene Symbol: Ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026275.4 | NP_080551.1 | 1, 2 | 223 | ||
| GenBank mRNA | AJ240086.1 | - | 1, 2 | 182 | ||
| AK003550.1 | - | 1, 2 | 243 | |||
| AK007517.1 | - | 1, 2 | 357 | |||
| AK075703.1 | - | 1, 2 | 233 | |||
| AK075714.1 | - | 1, 2 | 224 | |||
| BC011112.1 | - | 1, 2 | 231 | 3982808 | ||

Ube2r2
Genomic Map