siRNA Details
| Gene Symbol: Ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026275.4 | NP_080551.1 | 5 | 717 | ||
| GenBank mRNA | AJ240086.1 | - | 5 | 676 | ||
| AK003550.1 | - | 5 | 737 | |||
| AK007517.1 | - | 5 | 851 | |||
| AK075703.1 | - | 5 | 727 | |||
| AK075714.1 | - | 5 | 718 | |||
| BC011112.1 | - | 5 | 725 | 3982808 | ||

Ube2r2
Genomic Map