siRNA Details
| Gene Symbol: Mxra7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026280.3 | NP_080556.1 | 2, 3 | 380 | ||
| GenBank mRNA | AK012601.1 | - | 2, 3 | 352 | ||
| AK075801.1 | - | 2, 3 | 381 | |||
| BC075618.1 | - | 2, 3 | 347 | |||
| BC147320.1 | - | 2, 3 | 461 | 9056373 | ||
| BC147321.1 | - | 2, 3 | 461 | 9056376 | ||

Mxra7
Genomic Map