siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 12, 13 | 2869 | ||
| GenBank mRNA | AK016213.1 | - | 3, 4 | 391 | ||
| AK017665.1 | - | 1, 2 | 78 | |||
| AK156617.1 | - | 12, 13 | 2898 | |||
| BC052934.1 | - | 3, 4 | 474 | |||
| BC053435.1 | - | 12, 13 | 2869 | 6394578 | ||

Ctdp1
Genomic Map