siRNA Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026308.2 | NP_080584.1 | 1, 2 | 75 | ||
| GenBank mRNA | AJ504717.1 | - | 1, 2 | 85 | ||
| AK002780.1 | - | 1, 2 | 76 | |||
| AK003455.1 | - | 1, 2 | 73 | |||
| AY251016.1 | - | 1, 2 | 97 | |||
| BC028491.1 | - | 1, 2 | 85 | 1497187 | ||

Rpp21
Genomic Map