siRNA Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026313.1 | NP_080589.1 | 1, 2 | 223 | ||
| GenBank mRNA | AK014362.1 | - | 1, 2 | 216 | ||
| AK019464.1 | - | 1, 2 | 276 | |||
| AK078090.1 | - | 1, 2 | 270 | |||
| AK151228.1 | - | 1, 2 | 271 | |||
| AK151839.1 | - | 1, 2 | 271 | |||
| BC009092.1 | - | 1, 2 | 223 | 3157532 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108291, XM_006247155, XM_008768078, XM_006247156, FQ235074, and others.

Luc7l3
Genomic Map