siRNA Details
| Gene Symbol: 1700011L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026315.1 | NP_080591.1 | 1 | 230 | ||
| GenBank mRNA | AK005876.1 | - | 1 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700011L22Rik| Gene Symbol: 1700011L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026315.1 | NP_080591.1 | 1 | 230 | ||
| GenBank mRNA | AK005876.1 | - | 1 | 218 | ||