siRNA Details
| Gene Symbol: Stx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026343.2 | NP_080619.2 | 4 | 427 | ||
| GenBank mRNA | AK011400.1 | - | 4 | 421 | ||
| AK014718.1 | - | 3 | 366 | |||
| AK018526.1 | - | 4 | 417 | |||
| AK034718.1 | - | 4 | 416 | |||
| AK135240.1 | - | 4 | 423 | |||
| AK154503.1 | - | 4 | 428 | |||
| BC028639.1 | - | 4 | 414 | 1243532 | ||

Stx17
Genomic Map