siRNA Details
| Gene Symbol: Ttc39d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026351.2 | NP_080627.2 | 1 | 1445 | ||
| GenBank mRNA | AK016184.1 | - | 1 | 1028 | ||
| AK016869.1 | - | 1 | 892 | |||
| BC118946.1 | - | 1 | 1445 | 40092366 | ||
| BC118947.1 | - | 1 | 1445 | 40092368 | ||

Ttc39d
Genomic Map