siRNA Details
| Gene Symbol: 4930579F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163385.1 | NP_001156857.1 | 4 | 932 | ||
| NM_001163386.1 | NP_001156858.1 | 4 | 815 | |||
| GenBank mRNA | AK016318.1 | - | 4 | 815 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930579F01Rik| Gene Symbol: 4930579F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163385.1 | NP_001156857.1 | 4 | 932 | ||
| NM_001163386.1 | NP_001156858.1 | 4 | 815 | |||
| GenBank mRNA | AK016318.1 | - | 4 | 815 | ||