siRNA Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175121.3 | NP_780330.2 | 15 | 1710 | ||
| GenBank mRNA | AK031615.1 | - | 15 | 1741 | ||
| AK033812.1 | - | 3 | 250 | |||
| AK129342.1 | - | 15 | 1671 | |||
| AK164568.1 | - | 9 | 5473 | |||
| AK169378.1 | - | 5 | 469 | |||
| BC037675.1 | - | 12 | 1117 | |||
| BC041108.1 | - | 15 | 1580 | 4038546 | ||
| BC048178.1 | - | 15 | 1710 | 6825801 | ||
| BC049271.1 | - | 8 | 733 | |||
| BC057454.1 | - | 2 | 91 | |||

Slc38a2
Genomic Map