siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170479.1 | NP_001163950.1 | 3 | 365 | ||
| NM_026371.3 | NP_080647.2 | 3 | 507 | |||
| GenBank mRNA | AK007423.1 | - | 3 | 364 | ||
| AK018019.1 | - | 3 | 423 | |||
| AK089648.1 | - | 3 | 371 | |||
| AK151342.1 | - | 3 | 373 | |||
| AK169615.1 | - | 3 | 379 | |||
| BC023059.1 | - | 3 | 466 | 5355560 | ||
| DV652098.1 | - | 3 | 257 | |||
| DV658678.1 | - | 3 | 367 | |||

Loh12cr1
Genomic Map