siRNA Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026378.3 | NP_080654.3 | 8 | 1112 | ||
| GenBank mRNA | AK002439.1 | - | 8 | 1108 | ||
| AK003374.1 | - | 2 | 59 | |||
| AK149963.1 | - | 8 | 1113 | |||
| AK169405.1 | - | 8 | 1110 | |||
| AK171818.1 | - | 8 | 1113 | |||
| BC013483.1 | - | 8 | 1093 | 4242135 | ||
| BC058774.1 | - | 6 | 511 | |||

Dalrd3
Genomic Map