siRNA Details
Gene Symbol: Gm15063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042783.1 | - | 2 | 503 |
Gene Symbol: Rab39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175122.6 | NP_780331.1 | 2 | 567 | ||
GenBank mRNA | AB232636.1 | - | 2 | 343 | ||
AK018257.1 | - | 2 | 551 | |||
AK039013.1 | - | 2 | 541 | |||
AK048981.1 | - | 2 | 541 | |||
AK160737.1 | - | 2 | 541 | |||
BC050853.1 | - | 2 | 561 | 6517891 | ||
BC052472.1 | - | 2 | 492 | 6833461 |
