siRNA Details
| Gene Symbol: Rab39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175122.6 | NP_780331.1 | 2 | 804 | ||
| GenBank mRNA | AB232636.1 | - | 2 | 580 | ||
| AK018257.1 | - | 2 | 788 | |||
| AK039013.1 | - | 2 | 778 | |||
| AK048981.1 | - | 2 | 778 | |||
| AK160737.1 | - | 2 | 778 | |||
| BC050853.1 | - | 2 | 798 | 6517891 | ||
| BC052472.1 | - | 2 | 729 | 6833461 | ||

Rab39b
Genomic Map