siRNA Details
Gene Symbol: Atp1b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290389.1 | NP_001277318.1 | 8 | 1128 | ||
NM_133690.3 | NP_598451.1 | 8 | 1140 | |||
GenBank mRNA | AF348324.1 | - | 8 | 995 | ||
AF348325.1 | - | 8 | 983 | |||
AK053500.1 | - | 8 | 1079 | |||
BC137923.1 | - | 8 | 1081 | 8860942 | ||
BC137924.1 | - | 8 | 1093 | 8860943 |
