siRNA Details
| Gene Symbol: Cysrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026415.3 | NP_080691.2 | 2 | 354 | ||
| GenBank mRNA | AK003658.1 | - | 2 | 322 | ||
| AK009095.1 | - | 2 | 356 | |||
| BC117777.2 | - | 1 | 654 | 40091453 | ||
| BC117778.2 | - | 1 | 654 | 40091454 | ||

Cysrt1
Genomic Map