siRNA Details
| Gene Symbol: 2310033P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024210.2 | NP_077172.1 | 1 | 160 | ||
| GenBank mRNA | AK009601.1 | - | 1 | 147 | ||
| AK012628.1 | - | 1 | 171 | |||
| AK137066.1 | - | 198 | ||||
| BC002181.1 | - | 1 | 160 | 3487283 | ||

2310033P09Rik
Genomic Map