siRNA Details
| Gene Symbol: Camsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081360.1 | NP_001074829.1 | 13 | 3100 | ||
| GenBank mRNA | AK028542.1 | - | 11 | 3050 | ||
| AK122436.1 | - | 1 | 413 | |||
| BC100407.1 | - | 1 | 599 | |||
| BC138367.1 | - | 12 | 2676 | 8861387 | ||
| BC145258.1 | - | 11 | 2643 | 9053811 | ||

Camsap2
Genomic Map