siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 6 | 273 | ||
| GenBank mRNA | AK007788.1 | - | 6 | 237 | ||
| AK008280.1 | - | 6 | 236 | |||
| AK011737.1 | - | 6 | 253 | |||
| AK089948.1 | - | 6 | 239 | |||
| AK148146.1 | - | 6 | 243 | |||
| AK166332.1 | - | 6 | 267 | |||
| AK167301.1 | - | 6 | 242 | |||
| AK167321.1 | - | 6 | 237 | |||
| BC051554.1 | - | 5 | 237 | |||
| BC061065.1 | - | 6 | 272 | 6594260 | ||
| CJ075089.1 | - | 6 | 299 | |||

Ufm1
Genomic Map