siRNA Details
| Gene Symbol: Coa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174987.4 | NP_778152.1 | 1 | 206 | ||
| GenBank mRNA | AK008041.1 | - | 1 | 141 | ||
| AK009404.1 | - | 1 | 180 | |||
| BC024399.1 | - | 1 | 136 | 5026081 | ||
| BC038634.1 | - | 1 | 192 | 1513986 | ||
| BC046907.1 | - | 2 | 97 | 6394880 | ||
| BU562971.1 | - | 1 | 209 | |||
| CO039015.1 | - | 2 | 571 | |||

Coa6
Genomic Map