siRNA Details
Gene Symbol: Coa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174987.4 | NP_778152.1 | 1 | 293 | ||
GenBank mRNA | AK008041.1 | - | 1 | 228 | ||
AK009404.1 | - | 1 | 267 | |||
BC024399.1 | - | 1 | 223 | 5026081 | ||
BC038634.1 | - | 1 | 279 | 1513986 | ||
BC046907.1 | - | 2 | 184 | 6394880 | ||
BU562971.1 | - | 1 | 296 | |||
CO039015.1 | - | 2 | 484 |
