siRNA Details
| Gene Symbol: Ppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026438.4 | NP_080714.2 | 6, 7 | 608 | ||
| GenBank mRNA | AK008575.1 | - | 6, 7 | 609 | ||
| AK088397.1 | - | 6, 7 | 604 | |||
| AK145640.1 | - | 6, 7 | 658 | |||
| AK146300.1 | - | 6, 7 | 571 | |||
| AK151512.1 | - | 6, 7 | 599 | |||
| BC010468.1 | - | 6, 7 | 542 | 3585780 | ||
| CT010203.1 | - | 6, 7 | 501 | |||

Ppa1
Genomic Map