siRNA Details
| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026442.3 | NP_080718.1 | 3 | 330 | ||
| GenBank mRNA | AK002667.1 | - | 3 | 227 | ||
| AK008567.1 | - | 3 | 249 | |||
| AK011760.1 | - | 3 | 231 | |||
| AK020626.1 | - | 3 | 214 | |||
| AK168728.1 | - | 3 | 277 | |||
| BC048547.1 | - | 3 | 311 | |||
| BC056653.1 | - | 3 | 204 | 5068125 | ||
| CJ170455.1 | - | 3 | 331 | |||

Cmc1
Genomic Map