siRNA Details
| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026442.3 | NP_080718.1 | 2 | 203 | ||
| GenBank mRNA | AK002667.1 | - | 2 | 100 | ||
| AK008567.1 | - | 2 | 122 | |||
| AK011760.1 | - | 2 | 104 | |||
| AK020626.1 | - | 2 | 87 | |||
| AK168728.1 | - | 2 | 150 | |||
| BC048547.1 | - | 2 | 184 | |||
| BC056653.1 | - | 2 | 77 | 5068125 | ||
| CJ170455.1 | - | 2 | 204 | |||

Cmc1
Genomic Map