siRNA Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026445.2 | NP_080721.1 | 5 | 529 | ||
| GenBank mRNA | AK013017.1 | - | 5 | 509 | ||
| AK076022.1 | - | 5 | 535 | |||
| AK138300.1 | - | 5 | 520 | |||
| AK140169.1 | - | 6 | 544 | |||
| AK157784.1 | - | 5 | 530 | |||
| AK166216.1 | - | 5 | 508 | |||
| BC104404.1 | - | 3 | 638 | |||
| BC104405.1 | - | 3 | 638 | |||
| BC119479.1 | - | 3 | 638 | |||
| BC119480.2 | - | 3 | 638 | 40050522 | ||

Sumf2
Genomic Map