siRNA Details
| Gene Symbol: Coq9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026452.2 | NP_080728.1 | 8, 9 | 918 | ||
| GenBank mRNA | AK004527.1 | - | 4, 5 | 343 | ||
| AK009179.1 | - | 8, 9 | 930 | |||
| AK154343.1 | - | 8, 9 | 943 | |||
| AK158979.1 | - | 8, 9 | 937 | |||
| BC036386.1 | - | 8, 9 | 918 | 1067021 | ||

Coq9
Genomic Map