siRNA Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 10 | 662 | ||
GenBank mRNA | AK007549.1 | - | 10 | 633 | ||
AK007937.1 | - | 10 | 662 | |||
AK007960.1 | - | 10 | 652 | |||
AK010957.1 | - | 11 | 739 | |||
AK030594.1 | - | 642 | ||||
AK153481.1 | - | 10 | 663 | |||
BC006057.1 | - | 4 | 175 | |||
BC016117.1 | - | 11 | 944 | 4486610 |
Gene Symbol: Gm5434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136183.1 | - | 1 | 632 | ||
BC085248.1 | - | 1 | 638 |
