siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114977.1 | NP_001108449.1 | 17, 18 | 1906 | ||
| NM_026476.2 | NP_080752.2 | 16, 17 | 1774 | |||
| GenBank mRNA | AK029898.1 | - | 17, 18 | 1806 | ||
| AK161691.1 | - | 1, 2 | 151 | |||
| BC031497.1 | - | 17, 18 | 1766 | |||
| BC068265.1 | - | 16, 17 | 1790 | 30022109 | ||
| BC080699.1 | - | 16, 17 | 1655 | |||

U2surp
Genomic Map