siRNA Details
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114977.1 | NP_001108449.1 | 19 | 2035 | ||
NM_026476.2 | NP_080752.2 | 18 | 1903 | |||
GenBank mRNA | AK029898.1 | - | 19 | 1935 | ||
AK161691.1 | - | 2 | 2577 | |||
BC031497.1 | - | 19 | 1895 | |||
BC068265.1 | - | 18 | 1919 | 30022109 | ||
BC080699.1 | - | 18 | 1784 |
