siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114977.1 | NP_001108449.1 | 21, 22 | 2356 | ||
| NM_026476.2 | NP_080752.2 | 20, 21 | 2224 | |||
| GenBank mRNA | AK029898.1 | - | 21, 22 | 2256 | ||
| BC031497.1 | - | 21, 22 | 2225 | |||
| BC068265.1 | - | 20, 21 | 2240 | 30022109 | ||
| BC080699.1 | - | 20, 21 | 2105 | |||

U2surp
Genomic Map