siRNA Details
| Gene Symbol: Trabd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026485.2 | NP_080761.2 | 10 | 1127 | ||
| GenBank mRNA | AK005300.1 | - | 10 | 1108 | ||
| AK017745.1 | - | 10 | 1121 | |||
| AK131116.1 | - | 2 | 3209 | |||
| AK146027.1 | - | 10 | 1143 | |||
| BC005574.1 | - | 10 | 1153 | 3967961 | ||

Trabd
Genomic Map