siRNA Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026499.4 | NP_080775.3 | 3 | 467 | ||
| GenBank mRNA | AK004831.1 | - | 3 | 448 | ||
| AK012605.1 | - | 3 | 215 | |||
| AK051608.1 | - | 1 | 620 | |||
| AK133872.1 | - | 3 | 448 | |||
| AK146293.1 | - | 3 | 445 | |||
| AK159519.1 | - | 3 | 445 | |||
| AK171709.1 | - | 2 | 1731 | |||
| AK179065.1 | - | 2 | 96 | |||
| AK188277.1 | - | 2 | 76 | |||
| AK212807.1 | - | 1 | 52 | |||
| BC012039.1 | - | 3 | 454 | 2648111 | ||

Srsf6
Genomic Map