siRNA Details
| Gene Symbol: Fam122a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026520.3 | NP_080796.1 | 1 | 689 | ||
| GenBank mRNA | AK005219.1 | - | 1 | 679 | ||
| AK010777.1 | - | 1 | 700 | |||
| AK011741.1 | - | 1 | 673 | |||
| AK018219.1 | - | 1 | 641 | |||
| AK033646.1 | - | 1 | 689 | |||
| AK077654.1 | - | 1 | 671 | |||
| AK172089.1 | - | 1 | 674 | |||
| BC028637.1 | - | 1 | 657 | 1248203 | ||
| BC083062.1 | - | 1 | 679 | 5361132 | ||

Fam122a
Genomic Map