siRNA Details
| Gene Symbol: Rbm42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133693.2 | NP_598454.2 | 2 | 391 | ||
| GenBank mRNA | AK013929.1 | - | 2 | 329 | ||
| AK151000.1 | - | 2 | 303 | |||
| AK211257.1 | - | 1 | 44 | |||
| AK213653.1 | - | 1 | 44 | |||
| AK216265.1 | - | 1 | 44 | |||
| BC009148.1 | - | 2 | 328 | 3600406 | ||
| BC011286.1 | - | 2 | 310 | 3485410 | ||
| BC027372.1 | - | 2 | 315 | 4952304 | ||
| BC039917.1 | - | 2 | 326 | |||

Rbm42
Genomic Map