siRNA Details
| Gene Symbol: 2700062C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026529.4 | NP_080805.1 | 3, 4 | 324 | ||
| GenBank mRNA | AK011228.1 | - | 2, 3 | 411 | ||
| AK012467.1 | - | 2, 3 | 426 | |||
| AK013030.1 | - | 3, 4 | 232 | |||
| AK018391.1 | - | 3, 4 | 328 | |||
| AK164298.1 | - | 1, 2 | 2027 | |||
| BC037075.1 | - | 3, 4 | 372 | 5038294 | ||
| BC084681.1 | - | 3, 4 | 334 | 30144888 | ||

2700062C07Rik
Genomic Map